Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thermo Scientific® TSG Laboratory Refrigerators with Glass Door

Supplier: Thermo Fisher Scientific
Thermo Scientific®
TSG4505GA TSG1205GA TSG4505GD TSG5005GD TSG5005GA TSG3005GD TSG1205GA TSG3005GA TSG2305GA TSG4505GA TSG3005GA TSG1205GD TSG2305GD TSG3005GD TSG5005GD TSG2305GA TSG1205GD TSG4505GD TSG2305GD TSG5005GA
76764-976EA 10868.46 USD InStock
CA76764-964 76764-976 CA76764-976 76764-964 76764-978 CA76764-962 CA76764-940 CA76764-974 CA76764-952 76764-938 CA76764-978 CA76764-938 76764-980 76764-950 CA76764-950 76764-974 CA76764-980 76764-952 76764-962 76764-940
Thermo Scientific® TSG Laboratory Refrigerators with Glass Door
Refrigerators
Thermo Scientific TSG Series glass door laboratory refrigerators offer quiet, intuitive operation with excellent temperature performance and an outstanding warranty, meeting the needs of a wide variety of professional users who need consistent, reliable performance without sacrificing usability or sustainability.

  • Purpose-built from the ground up for clinical and research laboratory customers
  • Quiet operation, less than 53 dB
  • ENERGY STAR certified
  • Heat-free defrost
  • Insulated solid doors, self-closing door with 90° stop, keyed lock
  • Four 2" casters, front casters are lockable
  • Hi/Lo temperature, door ajar, and power failure alarms and remote alarm contacts
  • 4 years parts and labor and 1 year compressor warranty

All TSG Series laboratory refrigerators utilize SNAP and F-Gas compliant hydrocarbon refrigerants and are configured to provide excellent temperature performance while minimizing power consumption, all delivered from a zero waste to landfill facility.

Certifications: cULus, ENERGYSTAR.

Ordering information: Accessories available include chart recorders, independent temperature monitors, surge suppressors and more.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty