Would you like to log in again?
Thermo Scientific™ ART™ Pipette Tips
Supplier: Thermo Fisher Scientific
Thermo Scientific™ ART™
2069RC-HR 3551RC-HR 2065ERC-HR 3501RC-HR 2139RC-HR 2079RC-05-HR 3551RC-05-HR 2149PRC-HR 2179RC-HR 2065ERC-05-HR 3101RC-HR 2140RC-HR 3521RC-05-HR 2139RC-05-HR 3511RC-HR 3101RC-05-HR 2179RC-05-HR 2140RC-05-HR 3501RC-05-HR 2149PRC-05-HR 2069RC-05-HR 3511RC-05-HR 3521RC-HR 2079RC-HR
76809-080CS 772.5 USD InStock
76809-080 76809-060 76809-082 76809-070 76809-084 76809-062 76809-072 76809-064 76809-096 76809-074 76809-066 76809-110 76809-098 76809-076 76809-112 76809-068 76809-100 76809-078 76809-114 76809-102 76809-058 76809-104 76809-106 76809-108
Thermo Scientific™ ART™ Pipette Tips
Pipette Tips
Thermo Scientific Art™ pipette tip and rack mail-back recycling program provides a simple and effective solution for recycling single use plastics. The TipCycle recycle kit arrives with your case of pipette tips to collect used pipette tips and empty racks. Attach the pre-paid shipping label and return for recycling.
  • Recycle nonhazardous pipette tips, racks, and associated packaging with The Tipcycle Program. Sustainable design based on virgin resins
  • Guaranteed free of detectable DNA, DNase/RNase, endotoxins and heavy metals
  • Tested for endotoxin (pyrogen) contamination
  • Resins are pre-tested for metal contamination
  • Graduations allow a visual check of pipetting accuracy
Perform pipetting accurately and consistently with Thermo Scientific™ ART™ Barrier Hinged Rack Pipette Tips, which seal when exposed to potential contaminants. The pipette tips from the reload inserts clip securely into the environmentally friendly, reusable hinged racks and the covers may be used as hinged or lift-off style. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. ART hinged racks are available in Thermo Scientific™ MicroPoint™, extended length, low retention.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty