Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSMB10 Goat Polyclonal Antibody

Supplier: Antibodies.com LLC
A84682-100
77063-590EA 480 USD InStock
77063-590
Anti-PSMB10 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to PSMB10 for ELISA, WB and IHC with samples derived from Human.

  • Validated Applications: ELISA, WB, IH

Type: Primary
Antigen: PSMB10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty