Would you like to log in again?
Stainless Steel Tubing ¹/₁₆" OD
Supplier: Optimize Technologies
10-12-00165 10-12-00150 10-12-00160 10-12-00144 10-12-00152 10-12-00154 10-12-00145 10-12-00158 10-12-00148 10-12-00143 10-12-00163 10-12-00155 10-12-00161 10-12-00151 10-12-00149 10-12-00146 10-12-00162 10-12-00141 10-12-00156 10-12-00166
77100-090EA 207.59 USD InStock
77100-090 77100-092 77100-082 77100-094 10123-220 77100-114 77100-104 77100-116 77100-106 77100-084 77100-096 77100-086 77100-120 77100-098 77100-088 77100-110 77100-100 77100-122 77100-112 77100-102
Stainless Steel Tubing ¹/₁₆" OD
Tubing Chromatography Tubing
Pre-cut and micro polished tubes.
  • Seamless
  • Guaranteed square/burr-free ends
  • Passivated prior to shipment
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty