Would you like to log in again?
OPTI-LOK™ Low Pressure Fittings
Supplier: Optimize Technologies
OPTI-LOK™
10-21-00290 10-21-00283 10-21-00343 10-21-00296 10-21-00291 10-21-00310 10-21-00295 10-21-00318 10-21-00293 10-21-00326 10-21-00307 10-21-00312 10-21-00300 10-21-00305 10-21-00284 10-22-00319 10-21-00306 10-21-00308 10-22-00315 10-21-00281 10-21-00317 10-21-00299 10-21-00323 10-21-00313
21510-187EA 13.64 USD InStock
21510-187 21510-199 21510-201 21510-720 21510-776 77100-192 21510-722 77100-246 77100-224 77100-214 77100-248 77100-216 77100-206 77100-194 21510-712 21510-702 21510-714 77100-230 77100-198 77100-232 77100-210 77100-200 77100-222 21510-718
OPTI-LOK™ Low Pressure Fittings
Fittings and Connectors
OPTI-LOK LP Fittings are an excellent choice for making low-pressure connections into ¹/₄-28 ports. Connections to flat-bottomed ports can be made using OPTI-LOK flat-bottomed ferrules for ¹/₁₆ or ¹/₈" tubing without the need for pre-flanged tubing ends. OPTI-LOK LP Fittings are precision machined to deliver leak-free, zero-dead-volume connections every time. We also offer ¹/₄-28 unions in various through-hole sizes for joining flat-bottom fittings.
  • PEEK, ETFE, PTFE, PCTFE, black and white acetal nuts and ferrules for ¹/₈", 0.100" and ¹/₁₆" outer Ø tubing
  • Excellent choice for making low-pressure connections into ¹/₄-28 ports
  • Connections to flat bottomed ports can be made using OPTI-LOK® flat-bottomed ferrules
  • No need for pre-flanged tubing ends
  • Precision machined for optimal quality and thread consistency
  • Zero-dead-volume connections every time
We realize that your time is valuable and should not be wasted worrying about fittings. For this reason, all optimize fittings are precision machined in the USA for optimal quality and thread consistency. The machining process is the best way to ensure tight manufacturing tolerances and minimal product variance. What does all this mean? It means that every fitting you purchase from optimize will perform just as well as the last, so you'll have no worries about faulty fittings. This reflects our commitment to providing our customers with the highest quality, best designed product available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty