Would you like to log in again?
OEM Specific Fittings
Supplier: Optimize Technologies
40-24-01290 41-24-01281 40-24-01273 41-24-01280 41-24-01277 10-23-00358 40-24-01276 41-24-01278 10-20-00330 10-20-02875 40-24-01271 10-20-02878 41-24-01283 41-24-01282 10-20-00331 10-20-02877 40-24-01275 41-24-01279 10-20-02876 40-24-01291
77101-386EA 22.48 USD InStock
77101-386 77101-398 77101-384 77101-396 21510-762 77101-382 21510-764 77101-394 77101-380 77101-392 83008-160 83008-164 77101-388 77101-378 77101-400 83008-162 83008-158 89522-312 77101-390 21510-802
OEM Specific Fittings
Fittings and Connectors
OPTI-LOK Fittings are ideal for high-pressure connections with ¹/₁₆" OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.
  • OEM Specific Fittings (Nuts, Ferrules and Unions) for ¹/₁₆" Tubing
  • Waters, Valco, Parker and SSI styles available
  • Fittings are included with all Unions
  • Precision machined for optimal quality and thread consistency
  • Zero-dead-volume connections every time
We realize that your time is valuable and should not be wasted worrying about fittings. For this reason, all Optimize fittings are precision machined in the USA for optimal quality and thread consistency. The machining process is the best way to ensure tight manufacturing tolerances and minimal product variance. What does all this mean? It means that every fitting you purchase from Optimize will perform just as well as the last, so you will have no worries about faulty fittings. This reflects our commitment to providing our customers with the highest quality, best designed product available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty