Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Proteasome 20S LMP2 Rabbit Monoclonal Antibody [clone: ARC1629]

Supplier: Antibodies.com LLC
A308367-100 A308367-50
77229-658EA 305 USD InStock
77229-658 77229-656
Anti-Proteasome 20S LMP2 Rabbit Monoclonal Antibody [clone: ARC1629]
Antibodies
Rabbit monoclonal [ARC1629] antibody to Proteasome 20S LMP2 for WB and IHC with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200

Type: Primary
Antigen: Proteasome 20S LMP2
Clonality: Monoclonal
Clone: ARC1629
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty