The Cubis® II laboratory balances are modular, providing choice between applications and configurations that best suits the labratory needs. These balances can be configured at the level of display, draftshields, software applications and hardware functions. The Cubis® II range of high-capacity micro balances with a maximum load of between 32 and 111 g and a readability between 0.001 and 0.002 mg provide the ideal choice for a broad range of applications.
- Ultra-high resolution with up to 61 million weighing steps (digits)
- End-to-end data integrity. 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management
- Cleanability, part of compliance
- Error-free operation. Individual QApp workflows, motorised auto-leveling
- Allow upgrades for hardware features like automated draft shields or built-in ioniser
- Optional inner draft shield for best weighing performance
- Standard version, non verified
The Cubis® II Ultra-High Resolution Balances are precise analytical instruments used to weigh very small quantities of samples with high precision. The performance in a laboratory environment often deviates from instrument specifications due to varying environmental conditions and influences deriving from samples and users. The higher the resolution, the greater the relative measurement uncertainty is. Needless to say, these devices are particularly sensitive to imperfect laboratory conditions. The goal was to build a balance which is less prone to environmental effects, resulting in faster weighing workflows yet improved specifications.