Would you like to log in again?
MicroFunnel™ Disposable Filter Funnels
Supplier: Cytiva
MicroFunnel™
4801ME 4818 4804ME 4818 4800 4810 4828 4817 4815 4801ME 4815ME 4803 4819 4815 4819 4803 4800ME 4804 4817 4805 4800 4810 4815ME 4806 4810ME 4801 4852 4805 4828 4810ME 4818 4806 4800ME 4801 4818 4852 4804ME 4804
28143-542PK 361 USD InStock
28143-542 CA28143-574 CA28143-576 28143-544 CA28143-554 CA28143-556 77391-978 77391-974 CA77391-984 77391-972 77391-970 97003-750 43300-292 CA55095-060 83008-126 28143-560 CA28143-572 28143-540 CA28143-540 28143-576 28143-554 CA28143-542 28143-556 CA77391-978 CA28143-544 CA77391-974 CA97003-750 CA77391-972 CA77391-970 77391-984 55095-060 CA83008-126 CA43300-292 28143-572 28143-574 CA28143-560
MicroFunnel™ Disposable Filter Funnels
Funnels Membrane Filter Funnels
Save time with disposable units by eliminating the cleaning and sterilization required with reusable funnels.
  • Disposable
  • Ideal for aqueous solutions
  • Squeeze separation
MicroFunnel™ filter funnels are ideal for testing any aqueous solution for microbial contamination using the principles of Membrane Filter (MF) technique
The unique squeeze separation makes membrane retrieval easy. Choose from 0.45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications, 0.2 and 0.45 µm Supor® (polyethersulfone) membrane to minimize binding of inhibitory substances, or Metricel® Black (modified polyethersulfone) membrane for better contrast with light colored colonies. Available in 100 and 300 ml capacity. Graduations marked on outside of cylinder for better visibility and easy measurement. Gamma irradiated. Gamma irradiation dose is 15 to 30 kGy.
Applications include quality control analysis in pharmaceutical production, analyzing process water for industrial applications, and testing drinking water or wastewater for contamination.
Ordering information: Filter funnels are individually bagged except where indicated. Other types of Disposable Filter Funnels are available. See MicroFunnel™ ST Disposable Filter Funnels for a convenient and economical choice for sterility testing within isolators. See MicroFunnel™ Plus Disposable Filter Funnels for funnels designed specifically for water system monitoring—sample cup and filter funnel all in one. Sentino™ Microbiology Pump–designed specifically for MicroFunnel™ filter funnel. Contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty