Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guanidinium hydrochloride, Technical Grade low ash residue after ignition

Supplier: VWR
Warning
Synonyms: Guanidinium chlorideGuanidine hydrochlorideGuanidine chlorideAminoformamidine hydrochlorideGuanadine hydrochlorideAminomethanamidine hydrochlorideGuanidine HCI

VWRVE424-25KG E424-5KG E424-500G
77394-790EA 6054.6 USD InStock
77394-790 97062-750 97062-748
Guanidinium hydrochloride, Technical Grade low ash residue after ignition
Guanidinium hydrochloride
Denaturant that does not contain anti-caking agent.


Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 185 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg
Order Now
Specification Test Results
Ammonium Chloride 0.5%
Ash 0.4 %
Moisture (KF) 0.3 %
Purity 98.5 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty