Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultramark®

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: SpectraSynQ PFPE-1600

PC7953-1G
77433-480EA 21 USD InStock
77433-480
Ultramark®
Ultramark®
Fluorinated polyether for mass spectrometry up to mass 1960.


Boiling Pt: 128 °C/0.2 mm/Hg
Storage Temperature: Ambient
MDL Number: MFCD00166131
CAS Number: 66795-94-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty