Would you like to log in again?
Storage and Assay Microplates
Supplier: Agilent
204357-100 201240-100 203426-100 204600-100 204357-100 204600-100 203426-100 201240-100
77463-692PK 172 USD InStock
77463-692 77463-662 CA77463-682 77463-682 CA77463-692 CA77463-662 CA77463-702 77463-702
Storage and Assay Microplates
Microplates Deep Well Microplates
For biological assays, low storage temperatures, and filtrate collection applications.
  • Compatible with all major automation platforms and benchtop devices
  • No risk feasibility testing – test a sample in your application
  • Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
  • Molded with chemical resistant polymers, for maintained performance with prolonged exposure to aggressive solutions and solvents
  • Sealable with heat and adhesive seals, or cap mats, for superior performance
  • Low temperature tolerant for refrigerator or freezer storage
  • Autoclavable, can be sterilized at 121 °C, 15 psi for 20 minutes
  • Available in a large range of well volumes, from 58 µl to 10 ml for application scale-up or scale-down
  • Complementary fit to Agilent filter microplates for centrifugation stacking
  • Manufactured in a DNAse/RNase-free ISO 9001 regulated facility
Agilent supplies collection and storage assay microplates for a wide range of reaction chemistries, biological assays, low storage temperatures, and filtrate collection applications. These microplates are molded with medical-grade, chemical-resistant polymers and are available in 24, 48, 96, or 384-well configurations of variable volumes. Agilent also supplies mat seals and lids for these microplates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty