Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ProKlenz® ALIGN Neutral Pharmaceutical Detergent

Supplier: Steris
ProKlenz®
143305WR 143301WR
77530-438EA 2912.03 USD InStock
77530-438 77530-440
ProKlenz® ALIGN Neutral Pharmaceutical Detergent
Cleaning Agents Detergents and Surface Cleansers
ProKlenz® ALIGN neutral pharmaceutical detergent offers gentle but effective cleaning for a wide range of process residues in CIP and manual applications.

  • Neutral pH: Suitable for a wide array of manufacturing substrates and manual cleaning
  • Low foaming system: Supports use in high-impingement systems
  • Free rinsing neutral pH formula: Minimizes water and utility consumption with no neutralization required
  • Complete lot and change control traceability: Supports quality assurance and control
  • Specific and non-specific analytical methods: Indicates that chemical has been removed from contact surface and speeds up the validation process

Designed and developed specifically for pharmaceutical and oral dosage form manufacturers, ProKlenz ALIGN Detergent has broad substrate compatibility and a neutral pH that eliminates the need for wastewater neutralization. A robust technical documentation package supports validation.

Manufactured within an FDA registered plant, ISO 9001:2015 and 13485:2016 compliant. Meets the highest standard in manufacturing and quality assurance.

Applications: Manual cleaning, processing equipment cleaning: Process Vessels, reactors and blenders, component part cleaning and ultrasonic cleaning.

Foam: Low at temperatures above 40 °C
Solubility: Complete

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty