Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ecocool™ Refrigerated/Heating Circulating Baths

Supplier: Grant
ecocool™
ECOCOOL150RUS ECOCOOL100RUS
77531-688EA 5159 USD InStock
77531-688 77531-690
ecocool™ Refrigerated/Heating Circulating Baths
Circulating Thermostats Open Circulator Systems Heating/Cooling Circulators
A new range of innovative, eco-friendly, refrigerated and heating circulating baths offering significant running cost savings whilst delivering powerful cooling.

  • Pharmaceutical - mini pilot plant reactors
  • Food - refractometry
  • Life-science - electrophoresis cooling
  • Adjustable over temperature cut-out
  • Tank opening (h×w): 120×150 mm
  • High temperature cooling - active up to 150 °C
  • Min/max liquid level: ecocool 100R- 105/140 mm; ecocool 150R - 160/190 mm

Consisting of two models, all products in the ecocool range are supplied assembled as ready to use kits, complete with accessory hosing, clips and connectors as standard. Education - rotary evaporator cooling, replacement of running tap water cooling, immersing small samples, photometry, chromatography systems. Industrial - QC testing, sample preparation, general cooling, reaction chemistry, temperature control, semi-conductor manufacturing, rheometry.

Choice of two models, temperature range −30 to 150 °C (model dependent)
3 year warranty with Grant renowned service and support, no registration required
Active cooling through the whole temperature range
True energy saving of up to 80% against standard compressor units
Built in Britain to the highest specifications
Thermostat and chiller work in harmony, neither will operate alone, eliminating any danger of overheating or freezing
Single front switch for user convenience
Modern, sleek, attractive design

Delivery information: Assembled and supplied with standard tubing, insulation, clips and connectors.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty