Would you like to log in again?
TouchNTuff® 92-600 Disposable Nitrile Gloves with Enhanced Chemical Splash Protection
Supplier: Ansell
TouchNTuff®
92600110 105077 585835 9260000M-VEN 92600060 9260000L-VEN 9260011080 9260011090 9260011100 9260000XL-VEN 585836 585834 585837 9260000S-VEN
34800-500CS 425.15 USD InStock
34800-500 CA76006-390 CA76006-396 CA76006-392 CA76006-394 77587-114 77587-116 CA34800-506 CA34800-504 34800-506 CA34800-502 34800-504 CA34800-500 34800-502
TouchNTuff® 92-600 Disposable Nitrile Gloves with Enhanced Chemical Splash Protection
Gloves
An unparalleled combination of chemical splash protection, comfort, and durability.
  • TNT™ Technology for ultimate protection and advanced chemical resistance
  • Extra-soft, smooth feel, boosting comfort for extended wear
  • Proprietary nitrile resists rips and tears to provide outstanding durability
TouchNTuff® 92-600 gloves are the world's leading disposable gloves for chemical splash protection. Made with TNT™ chemical splash resistance technology, these gloves effectively safeguard hands from exposure to harmful chemicals, with a super-soft nitrile consturction that provides a comfortable feel during extended wear. Proprietary formulation is extra-strong to prevent tearing during heavy-duty work. The silicone-free design prevents contaminant transfer that causes flaws in paints, coatings, and finishes.
Running on 59% renewable energy recycling min. 14% of water to reduce freshwater usage. Certified by Intertek for diverting >99% of waste from landfill. With accredited environmental management system. Plastic-free, recyclable packaging includes min. 70% recycled material.
Certifications: Category III. EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016. FDA21 CFR 177-2600. US Food Contact Approved. ISO 18889:2019, ISO 9001, UKCA.
Caution: Do not use against heat or cold. Do not use for mechanical nor extended chemical protection.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty