Would you like to log in again?
DuPont™ Tyvek® IsoClean® Coveralls with Raglan Sleeves
Supplier: DuPont
Tyvek® IsoClean®
IC182BWHLG002500 IC182BWHSM002500 IC182BWHMD00250C IC182BWH3X00250C IC182BWHMD002500 IC182BWH4X00250C IC182BWHSM00250C IC182BWHMD0025CS IC182BWHLG002500 IC182BWH2X002500 IC182BWHMD0025CS IC182BWHXL00250C IC182BWHMD002500 IC182BWH3X00250C IC182BWH3X0025CS IC182BWH3X00250C IC182BWH2X00250C IC182BWHXL00250C IC182BWHXL002500 IC182BWHSM00250C IC182BWHLG00250C IC182BWH4X00250C IC182BWH4X00250C IC182BWH4X002500 IC182BWHXL0025CS IC182BWHSM00 IC182BWHMD00250C IC182BWH3X002500 IC182BWHSM002500 IC182BWH3X0025CS IC182BWH2X0025CS IC182BWHXL0025CS IC182BWHSM0025CS IC182BWH4X0025CS IC182BWH2X00250C IC182BWHXL002500 IC182BWHLG0025CS IC182BWHLG0025CS IC182BWH3X002500 IC182BWH2X0025CS IC182BWHLG00250C IC182BWH4X0025CS IC182BWH2X002500
80076-842CS 1032.02 USD InStock
CA80076-778 CA80076-836 CA80077-280 80076-842 CA80076-774 80076-844 CA80076-776 80077-274 CA80077-284 80077-278 CA80076-838 80077-078 80077-276 89129-638 CA80077-278 89129-636 89129-634 80076-772 CA80076-782 CA80076-784 80076-774 80076-836 CA80076-844 80076-838 80076-776 CA80076-840 80077-282 80076-778 CA80076-842 80077-280 89129-630 80077-284 CA80077-078 80077-048 CA89129-638 89129-628 CA80077-048 CA89129-632 80076-784 80076-840 CA80076-772 10016-072 80076-782
DuPont™ Tyvek® IsoClean® Coveralls with Raglan Sleeves
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Bound seams
  • Bound neck
  • Raglan sleeve design
  • Covered elastic wrists and ankles
  • Zipper closure
Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration. Coveralls have elastic openings at wrists and ankles for a tighter fit. Bound neck ensures lower particle shedding. Front zipper closure allows for easy donning and doffing. Raglan sleeve design allows for greater range of motion and comfort.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Order Now
ORDER
SPECIFICATIONS
COMPLEMENTARY PRODUCTS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty