Would you like to log in again?
Solujet® Low-Foaming Phosphate-Free Liquid Detergents
Supplier: Alconox
Solujet®
2155 2101 2115 2105 2130 2105 2115 2130 2101 2155
80077-359CS 440.41 USD InStock
80077-359 CA80077-366 CA80077-359 80077-362 CA80077-360 80077-360 80077-366 CA80077-364 80077-364 CA80077-362
Solujet® Low-Foaming Phosphate-Free Liquid Detergents
Cleaning Agents Detergents and Surface Cleansers
Non ionic, low foaming phosphate free liquid cleaner for use in labware washers, manual and ultrasonic cleaning. Corrosion inhibited formula recommended for glassware, metals, plastic, ceramic, porcelain, rubber and fiberglass. Free rinsing for reliable results and no interfering residues. Passes inhibitory residue test.
  • Concentrated, biodegradable and readily disposable
  • Low-foaming high-wetting detergent for maximum performance
  • Ideal for CIP cleaning and floor standing lab washing machines
  • Dilute 1:200
For removeing soil, radioisotopes, radioactivity, organics, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents.
Recommended for glass, stainless steel, PTFE, ceramic, cement and alkali resistant plastics and metals such as stainless steel. Solujet is inhibited for Aluminum. Clean-in place, laboratory warewashers, dishwashers, washer-sterilizers, washer-sanitizers, cart washers, spray washers, electrocleaning, ultrasonic cleaners, and manual cleaning if proper skin and eye protection is used.
Note: Corrosion testing is advisable.
They can be used to clean pharmaceutical process equipment, water testing labware, phosphate sensitive lab equipment cleaning, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors.
Certifications: USDA authorized category A1 for use in food contact surface cleaning. FDA certified.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty