Would you like to log in again?
Axygen® Robotic Pipette Tips, PK Series, Corning
Supplier: Corning
Axygen®
PK-384-R PK-384-R-S PKF-384-R-S
80086-038CS 3908.17 USD InStock
80086-038 80086-040 80086-042
Axygen® Robotic Pipette Tips, PK Series, Corning
Pipette Tips
The PK series robotic tips for the PerkinElmer® and Packard® Systems are certified RNase-, DNase-, and pyrogen-free.
The PK series robotic tips are engineered to exact dimensions of OEM tips to ensure automation compatibility.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty