Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitric acid 67 - 70%, Environmental Grade

Supplier: SEASTAR CHEMICALS
Danger
Synonyms: Nitric acid solution

62764-320 62764-460 62764-460
CA11021-138EA 464.11 USD InStock
CA11021-138 80722-306 CA71007-854
Nitric acid 67 - 70%, Environmental Grade
Nitric acid


Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608
Order Now
Specification Test Results
Assay 67 - 70%
Colour (APHA) 10 max.
Aluminum (Al) (10) 1 ppb max.
Antimony (Sb) 0.5 ppb max.
Arsenic (As) 0.5 ppb max.
Barium(Ba) 0.1 ppb max.
Beryllium (Be) 0.1 ppb max.
Bismuth (Bi 0.1 ppb max.
Boron (B) (5) 1 ppb max.
Cadmium (Cd) 0.5 ppb max.
Calcium (Ca) (50) 1 ppb max.
Cerium (Ce) 0.1 ppb max.
Cesium (Cs) 0.1 ppb max.
Chromium (Cr) 1 ppb max.
Cobalt (Co) 0.5 ppb max.
Copper (Cu) 0.5 ppb max.
Dysprosium (Dy) 0.1 ppb max.
Erbium (Er) 0.1 ppb max.
Europium (Eu) 0.1 ppb max.
Gadolinium (Gd) 0.1 ppb max.
Gallium(Ga) 0.1 ppb max.
Germanium(Ge) 0.1 ppb max.
Gold(Au) 0.1 ppb max.
Hafnium (Hf) 0.1 ppb max.
Holmium (Ho) 0.1 ppb max.
Indium (In) 0.1 ppb max.
Iron (Fe) (5) 1 ppb max.
Lanthanum (La) 0.1 ppb max.
Lead (Pb) 0.1 ppb max.
Lithium (Li) 0.1 ppb max.
Lutetium (Lu) 0.1 ppb max.
Magnesium (Mg) (5) 1 ppb max.
Manganese (Mn) 0.1 ppb max.
Mercury (Hg) 0.1 ppb max.
Molybdenum (Mo) 0.1 ppb max.
Neodymium (Nd) 0.1 ppb max.
Nickel (Ni) 0.5 ppb max.
Niobium (Nb) 0.1 ppb max.
Palladium (Pd) 0.5 ppb max.
Platinum (Pt) 0.5 ppb max.
Potassium (K) (5) 1 ppb max.
Praseodymium (Pr) 0.1 ppb max.
Rhenium (Re) 0.1 ppb max.
Rhodium(Rh) 0.5 ppb max.
Rubidium (Rb) 0.1 ppb max.
Ruthenium (Ru) 0.5 ppb max.
Samarium (Sm) 0.1 ppb max.
Scandium (Sc) 0.1 ppb max.
Selenium (Se) 1 ppb max.
Silver (Ag) 0.1 ppb max.
Sodium (Na) (50) 1 ppb max.
Strontium (Sr) 0.1 ppb max.
Tantalum (Ta) info only
Tellurium (Te) 0.1 ppb max.
Terbium (Tb) 0.1 ppb max.
Thallium (Tl) 0.1 ppb max.
Thorium (Th) 0.1 ppb max.
Thulium (Tm) 0.1 ppb max.
Tin (Sn) 0.5 ppb max.
Titanium (Ti) 0.5 ppb max.
Tungsten (W) 0.1 ppb max.
Uranium (U) 0.1 ppb max.
Vanadium (V) 0.5 ppb max.
Ytterbium (Yb) 0.1 ppb max.
Yttrium (Y) 0.1 ppb max.
Zinc (Zn) (2) 0.5 ppb max.
Zirconium (Zr) 0.1 ppb max.
Maximums stated applicable at time of bottling. Values in brackets are likely maximum values after storage in glass.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty