Would you like to log in again?
Monoject™ Tuberculin Syringes, Sterile, Covidien
Supplier: Covidien
Monoject™
8881511235 1180127012 1180128012 1180100777 1180125058 8881501160 1180126038 1180100555 8881500105 8881501400 8881501178 8881501368 1180125158 1180100555
82002-232CS 208.97 USD InStock
82002-232 82002-234 MJ8881-501178 53498-486 CA82002-232 82005-720 82002-240 82002-242 MJ8881-501368 MJ8881-500105 95029-584 82002-236 MJ8881-501160 82002-238
Monoject™ Tuberculin Syringes, Sterile, Covidien
Syringes Syringes, Medical
These latex-free syringes feature an ACCU-TIP flat plunger tip and ACCU-SCALE graduations of 0.01mL
Syringes are also available packaged individually in RigidPacks or SoftPacks. RigidPacks feature a durable, clear, plastic case with a twist-off top. SoftPack syringes feature a one-step peel-away package that facilitates quick and easy opening for aseptic handling. Safety syringes are equipped with a two-step locking mechanism shield that engages without splatter and can be safely locked with fluid still in barrel. All syringes are 1mL in volume, except for MJ8881-500105 and 87002-276, which are 0.5mL.
The clear barrel construction allows precise medication measurement and easier visualization of flashback. Syringes are available with permanently attached or detachable ultra-comfort, anti-coring needle. Syringes with permanently attached needles have low dead space.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty