Would you like to log in again?
KleenGuard™ A40 Liquid and Particle Protection Coveralls
Supplier: Kimberly-Clark
KleenGuard®
44307 44322 44317 44305 44312 44302 44315 44332 44314 30904 44327 44332 44306 44326 44316 44334 44333 44326 44335 44313 44314 44305 44323 44307 44336 44324 44304 44317 44315 44337 44303 44333 44316 44306 44303 44327 44325 44324 44337 44334 44335 44336 44325 44313 44312 44302 44304 44323
82007-564CS 216.61 USD InStock
82007-564 82007-586 CA82007-594 CA82007-572 82007-562 82007-584 CA82007-592 CA82007-570 82007-568 CA82007-576 CA82007-598 82007-600 82007-566 82007-588 CA82007-574 CA82007-596 CA82007-558 82007-604 CA82007-578 82007-602 CA82007-580 82007-570 82007-592 82007-590 CA82007-584 CA82007-562 82007-596 82007-574 CA82007-582 CA82007-560 82007-594 82007-572 CA82007-566 CA82007-588 82007-578 CA82007-564 CA82007-586 82007-576 82007-598 CA82007-602 CA82007-568 CA82007-600 82007-558 CA82007-604 77552-876 82007-560 CA82007-590 82007-580
KleenGuard™ A40 Liquid and Particle Protection Coveralls
Disposable Garments
KleenGuard™ A40 Coveralls are designed to be used in applications where Tyvek® garments are typically worn, and provide an excellent particle barrier and resist nonhazardous liquids.
  • Zippered front
  • Breathable white microporous film keeps out debris, non-hazardous liquid chemicals and dry particulates
  • Provides a better liquid barrier and a better particulate barrier than DuPont Tyvek®
  • Pass NFPA 99 for antistatic materials
  • Pass ASTM F1670 testing for penetration of blood and bodily fluids
  • Meet the ANSI/ISEA 101-1996 sizing standards
KleenGuard™ A40 liquid and particle protection coveralls feature a zippered front. Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates, the KleenGuard™ A40 material provides employees with a better liquid barrier and a better particulate barrier than DuPont Tyvek®.
Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing, and many more.
Certifications: Pass NFPA 99 for antistatic materials; pass ASTM F1670 testing for penetration of blood and bodily fluids; and meet the ANSI/ISEA 101-1996 sizing standards.
Ordering information: Coveralls 89005-054 and -056 are individually wrapped.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty