Would you like to log in again?
Hybridization Buffers, G-Biosciences
Supplier: G-Biosciences
786-471 786-538 786-470 786-024 786-538 R017 786-559 786-470 786-558 R015 R015 786-023 786-471 786-558 786-024 R016 R018 786-525 R016 786-023 R018 786-559 R017 786-525
82021-476EA 82.9 USD InStock
CA82021-482 82021-476 CA95043-538 CA82021-476 82021-478 95043-538 CA82021-478 82021-480 82021-482 CA82021-480 CA89167-768 CA89167-712 95029-246 89167-712 89167-768 CA89167-710 95029-248 89167-710 CA95029-246 CA95029-248 CA95043-512 CA89167-770 89167-770 95043-512
Hybridization Buffers, G-Biosciences
Buffers Electrophoresis Buffers Hybridization Buffers
G-Biosciences offers various sized supplies of the following hybridization buffers: Church & Gilbert’s [2X], Denhardt Solution [100X], Hybridization Denaturing Solution, Hybridization Neutralizing Solution, SSC [20X], SSPE [20X], TMAC Hyb Solution [3X], and TNT Buffer.
G-Biosciences' Church & Gilbert’s Hybridization Buffer is prepared from molecular biology grade SDS (sodium dodecyl sulfate), monosodium phosphate (NaHPO₄), EDTA (ethylenediaminetetracetic acid) and bovine serum albumin (BSA) Fraction V using DNase-, RNase- and Protease-free water. It consists of 1mM EDTA, 1% BSA, 0.5M sodium phosphate monobasic, and 7% SDS (at a pH of 7.2).
G-Biosciences' Denhardt Solution is suitable for nucleic acid hybridization. Denhardt's solution is a mixture of high-molecular weight polymers capable of saturating non-specific binding sites and artificially increasing the concentration of available probe. It consists of 2% Ficoll 400, 300mM NaCl, 2% polyvinylpyrrolidone, and 2% BSA.
The SSC or Saline-Sodium Citrate Buffer is a hybridization buffer to control stringency for washing steps in protocols for Southern blotting, in situ hybridization, DNA Microarray or Northern blotting. It consists of 0.3 M sodium citrate (at a pH of 7.0) and 3 M NaCl.
The SSPE or or Saline-Sodium phosphate-EDTA Buffer is a general hybridization buffer that consists of 0.2 M phosphate buffer (at a pH of 7.4), 2.98 M NaCl, and 0.02 M EDTA.
The TMAC Hybridization Solution consists of 3 M Tetramethyl ammonium chloride, 50mm Tris-HCl (at a pH of 8.0), 1 mm EDTA (at a pH of 8.0), and 0.1% SDS.
The TNT Buffer is a general molecular biology and hybridization buffer that consists of 0.1 M Tris.HCl (at a pH of 7.5), 0.15 M NaCl, and 0.05% Tween®-20.
Ordering information: Accessories available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty