Would you like to log in again?
OPTI-LYNX™ Guard Column Cartridges
Supplier: Optimize Technologies
OPTI-LYNX™
11-03782-EB 11-03782-JD 11-03128-EA 11-02630-JD 11-03782-EG 11-03128-EE 11-02630-EH 11-03128-ED 11-02630-EF 11-02630-EB 11-03128-EH 11-03782-EF 11-02630-EE 11-03128-EB 11-02630-EG 11-02630-EC 11-03782-EC 11-03782-ED 11-02630-ED 11-02630-EA 11-03782-EH 11-03782-EE 11-03128-EG 11-03128-EF 11-03128-JD 11-03128-EC 11-03782-EA
83008-192PK 1047.31 USD InStock
83008-192 89207-752 89207-730 89207-750 83008-196 83008-194 83008-200 89207-726 89207-748 89207-746 83008-204 89207-744 83008-202 89207-742 83008-206 89207-728 89207-762 89207-740 89207-760 89207-738 83008-198 89207-758 89207-736 89207-756 89207-734 89207-732 89207-754
OPTI-LYNX™ Guard Column Cartridges
Chromatography Columns
The OPTI-LYNX™ guard column is a high performance, cartridge-based guard system, suitable for use with any analytical column.
  • Can be used with any brand of column
  • Cartridge based system
  • Low dispersion cartridge interface
  • ZDV holder connection
  • Less than 2 µl swept volume
  • Extends column life
  • Provides effective protection with negligible impact
  • Rated to 6,000 psi
Some cartridge-based guard column products offer convenience with a price: higher internal volumes. At Optimize Technologies, we refuse to compromise performance for anything. OPTI-LYNX™ utilizes low dispersion component interfaces to keep extra column volume to a minimum: connection paths have a total internal volume of less than 2 µl. You get quick-connect convenience and low dispersion, all in one package.
Ordering information: Available individually and in packs of five. Custom packing available.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty