Would you like to log in again?
KIMAX® Separatory Funnel, Globe, [ST] Glass Stopper, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
29043-250
89002-082CS 254.14 USD InStock
89002-082
KIMAX® Separatory Funnel, Globe, [ST] Glass Stopper, Kimble Chase, DWK Life Sciences
Funnels Separating Funnels
Funnel is supplied with a [ST] ground glass stopcock and stopper
Sharp separations are achieved with a narrow bore in the body, just above the stopcock. Gradually-tapering shoulders and a wide neck facilitate cleaning.
Certifications: Designed from ASTM Specification E1096, Type 2 requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty