Would you like to log in again?
ART® and Hydrologix (HLT) SoftFit-L™ Pipette Tips for Rainin® LTS Pipettors, Molecular BioProducts
Supplier: Thermo Scientific ART
ART® SoftFit-L™
3781-RI 3731-05-RI 3752-05-HR 3722-HR 3722-RI 2779-HR 3721-RI 3732-05-HR 3731-05-HR 2769-05-HR 2769-HR 2789-HR 3751-RI 3751-05-RI 3732-RI 3751-05-HR 3721-05-HR 3721-HR 3742-05-RI 3706-ER 3781-05-RI 2789-05-HR 3752-HR 2749-HR 3731-HR 3741-HR 3742-HR 3752-05-RI 3752-RI 3781-05-HR 3704-ER 3721-05-RI 2739-05-HR 3741-RI 2779-05-HR 3722-05-HR 2739-HR 3732-HR 3751-HR 3731-RI 3782-05-RI 3782-HR 3742-RI 3722-05-RI 3732-05-RI 3781-HR 3782-05-HR 3782-RI 2749-05-HR
89031-422CS 1110.39 USD InStock
89031-422 89031-424 89031-386 89031-388 89031-420 89031-426 89031-428 89031-382 89031-384 89031-380 89031-434 89031-356 89031-436 89031-358 89031-430 89031-396 89031-352 89031-432 89031-398 89031-354 89031-438 89031-392 89031-350 89031-394 89031-390 89031-368 89031-400 89031-402 89031-446 89031-364 89031-366 89031-442 89031-408 89031-404 89031-406 89031-360 89031-362 89031-412 89031-378 89031-414 89031-452 89031-374 89031-410 89031-376 89031-416 89031-418 89031-370 89031-450 89031-372
ART® and Hydrologix (HLT) SoftFit-L™ Pipette Tips for Rainin® LTS Pipettors, Molecular BioProducts
Pipette Tips
SoftFit-L™ pipette tips are available in five volume sizes, and are designed for use with Rainin® LTS pipettors.
  • Certified pure
  • Reload inserts are color-coded by size
Tips deliver highly accurate samples, seal well on the pipettor, and are easy to seat and eject. To accommodate a variety of applications, SoftFit-L™ pipette tips offer an expanded selection of ART®, HLT, and low retention designs. The ART® (Aerosol Resistant Tips) series features a self-sealing barrier designed to prevent contamination even under the most demanding conditions.
These tips guarantee 100% protection against aerosol and liquid contamination. Low retention tips are designed to reduce sample retention. Molded from virgin polypropylene, they do not use additives or coatings that may affect sample integrity. Low retention tips ensure reduced sample loss due to adhesion, allowing for more accurate dispensing and an increased level of reproducibility from one procedure to the next.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty