Would you like to log in again?
Bottom Drip Adapter for Big Columns, Two Filters, Ace Glass Incorporated
Supplier: Ace Glass
5857-86 5857-89
89053-040EA 584.56 USD InStock
89053-040 89053-038
Bottom Drip Adapter for Big Columns, Two Filters, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This end-fitting adapter fits large columns with #80 Ace-Thred connections.
  • Recessed
  • Two polyethylene filter discs
  • Bottom outlet thread accepts 1/2-inch or 3/8-inch female NPT fitting
Adapter is recessed to allow the perforated support plate and retaining ring to fit. Bottom outlet is a thread that will accept a 1/2-inch or 3/8-inch female NPT fitting.
Ordering information: Complete item includes adapter, size -235 FETFE o-ring, 89052-914 perforated glass support plate, PTFE (89053-020) retaining ring, 100 and 350 micron polyethylene filter discs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty