Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
Electromagnetic Coil, Large Distilling Heads, Ace Glass
Supplier: Ace Glass
6592-10
89055-678EA 701.83 USD InStock
89055-678
Electromagnetic Coil, Large Distilling Heads, Ace Glass
Magnets
Designed to work with large distilling heads, this electromagnetic coil has a high permeability core to operate a takeoff device on standard automatic distilling heads.
It is fastened to the head via a special clamp (comes complete with clamp and wires to connect to automatic time switch)
The apparatus is 76 mm O.D. by 44 mm high; the diameter of the metal slug is 28.6 mm.
For intermittent service only; 30 seconds maximum ON, 25% maximum duty cycle.
The unit is designed for operation at 120 volt, 50/60 Hz AC maximum, or at 24 volts DC maximum.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty