Would you like to log in again?
KLEENGUARD® A70 Chemical Spray Protection Smocks, Kimberly-Clark Professional®
Supplier: Kimberly-Clark
KLEENGUARD®
09829 09829 09830 09830
89065-964CS 450.25 USD InStock
CA89065-962 89065-964 CA89065-964 89065-962
KLEENGUARD® A70 Chemical Spray Protection Smocks, Kimberly-Clark Professional®
Disposable Garments
These chemical spray protection smocks are constructed of a 1.5mil polyethylene film-coated, spunbond polypropylene fabric.
  • Seamless design
  • Soft and durable fabric, and a wick-away lining for increased comfort
  • Elastic wrists, tie front, and open back
  • NFPA 99 compliant for antistatic material
The KLEENGUARD® A70 line of apparel is designed to provide protection against chemical spray in hazardous waste remediation, environmental cleanup, petrochemical/oil refining, acid/caustic handling, biohazard cleanup, and pharmaceutical manufacturing applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty