Would you like to log in again?
LabKlenz™ 200 Acid Detergents
Supplier: Steris
LabKlenz™
1L2002 1L2005 1L2008 1L2008
89089-208EA 1784.61 USD InStock
89089-208 CA89089-212 89089-212 89089-210
LabKlenz™ 200 Acid Detergents
Cleaning Agents Detergents and Surface Cleansers
LabKlenz™ 200 Detergent is a low-foaming, phosphoric acid based liquid detergent.
  • Flexibility - Effective removal of laboratory residues in both soft and hard water
  • Cost Savings - Reduces utility consumption and cleaning time due with low foaming, free-rinsing formulation
  • Environmentally Friendly - Formulated with biodegradable surfactants
  • Reliability - Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility
LabKlenz™ 200 Detergent is specially formulated for use in automated glassware washing applications both as a rinse aid and for the removal of particulate, scale, and inorganic soils.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty