Would you like to log in again?
KIMAX® KimCote® Squibb Separatory Funnels with Glass Stopper, DWK Life Sciences
Supplier: DWK Life Sciences
KIMAX®
KC29048T-125 KC29048T-500 KC29048T-1000 KC29048T-250 KC29048T-2000
89097-436CS 658.91 USD InStock
89097-436 89097-434 89097-432 89097-430 89097-438
KIMAX® KimCote® Squibb Separatory Funnels with Glass Stopper, DWK Life Sciences
Funnels Separating Funnels
KIMAX® KimCote® plastic-coated, pear-shaped funnel.
Lower stems have an I-Ø large enough so that a column of liquid will 'break' with the stopcock closed, thereby giving a more complete separation, and eliminating the necessity of emptying the funnel.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Certifications: Meets ASTM Specification E1096, Type IV requirements.
Delivery information: Supplied with a [ST] ground glass stopper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty