Would you like to log in again?
ProKlenz® NpH Neutral Detergents
Supplier: Steris
ProKlenz®
143001WR 143005WR
89130-316EA 350.83 USD InStock
89130-316 89130-318
ProKlenz® NpH Neutral Detergents
Cleaning Agents Detergents and Surface Cleansers
ProKlenz® NpH Neutral Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms.This low foaming product removes a wide range of process residues and is ideal for use in CIP, COP, and manual applications.
  • Efficacy - Neutral formulation delivers excellent cleaning characteristics and substrate compatibility with enhanced operator safety
  • Easy to validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Cost savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics when used in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • Globally availability - phosphate free, and manufactured with biodegradable ingredients to support harmonization of cleaning processes
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
ProKlenz® NpH High performance neutral detergent is specially formulated to deliver effective cleaning with excellent substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty