Would you like to log in again?
ADPTR CS4
Supplier: 3M OCCUPATIONAL HEALTH
Adapter^ This adapter is designed to adapt the v-100, v-200, v-300 and v-400 air regulating valves to accommodate the bt-200 and bt-222 breathing tubes.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADPTR CS4 89132-102 7000002156 Case of 4 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty