Would you like to log in again?
uTIP™ Filtered Pipette Tips for Universal Pipettors, Biotix
Supplier: Biotix
uTIP™
63300045 63300043 63300047 63300153 63300041 63300044 63300046 63300154 63300042
89136-580CS 804.08 USD InStock
89136-580 76175-406 89136-582 76175-412 89136-572 89425-662 89136-576 89136-568 89408-550
uTIP™ Filtered Pipette Tips for Universal Pipettors, Biotix
Pipette Tips
Biotix filtered tips provide protection against aerosol contamination (cross contamination) with a 99.99% bacterial filter efficiency (BFE). uTIP is compatible with major brands like Eppendorf, Gilson, Biohit and Thermofisher.
  • FlexFit® technology features a flexible proximal end to ensure a secure seal with less force then competitor tips
  • X-Resin® is our proprietary low retention resin which maximizes sample recovery and improves accuracy and precision
  • Blade® Technology makes a fine point at the distal end of the tip ensuring a full dispense without the need for tip touch off
Each lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled and inspected to ensure only the best go into our products. All racks include a label with a unique bar-coded serial number, part number, and description for traceability.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty