Would you like to log in again?
IgM (mouse), ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ADI-900-120
89141-174EA 477 USD InStock
89141-174
IgM (mouse), ELISA Kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of IgM in ascites fluid, culture supernatants, and serum of mouse origin.
The IgM (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 90 minutes. Absorbance is read at 450nm. Ideal for determining the isotype and concentration of monoclonal antibodies.
Ordering information: Contains GxM IgG Microtiter plate, mouse IgM conjugate, Assay buffer 13 concentrate, Wash buffer concentrate, mouse IgGM standard, TMB Substrate, Stop solution 2.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty