Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD40 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
ADI-CSS-180-D ADI-CSS-180-F
89141-830EA 559 USD InStock
89141-830 89141-828
Anti-CD40 Rabbit Polyclonal Antibody
Antibodies
CD40 is a 45-50 kDa type I membrane protein and a member of the tumor necrosis factor (TNF) receptor superfamily. CD40 is also known as pb50 or CDW40.  CD40 is expressed primarily by professional antigen-presenting cells and it is also expressed on non-immune cell types. The CD40 expressed by professional antigen-presenting cells plays a critical role in co-stimulation and antigen-presenting cell activation in T cell-dependent immune responses. Signals generated through CD40 in B cells are anti-apoptotic. These signals are also required for T cell-dependent B cell activation and proliferation, isotype switching, up-regulation of costimulatory receptors, germinal center formation and memory generation. CD40 signal transduction is initiated by binding trimeric CD40 ligand (CD40L) which is found on the surface of activated T cells.  CD40 engages several signalling pathways in B cells and these include NF-kB, the mitogen-activated protein kinases p38 and c-Jun N-terminal kinase. The 62 amino acid cytoplasmic domain (CD40c) of CD40 contains two linear TNF-receptor associated factor (TRAF) binding domains. The membrane proximal site of CD40c binds TRAF6 and the membrane distal site binds TRAF1, 2 and 3. CD40c is thought to mediate the CD40-dependant signalling pathways by recruitment of TRAF to this multimerized domain. CD40 ligation has also been shown to stimulate iNOS expression, and therefore NO production, by activating NF-kB in IFN-g treated microglial cells. NO derived from microglia has been implicated in the damage of myelin-producing oligodendrocytes in demyelinating disorders like multiple sclerosis (MS) and neuronal death during Alzheimer’s disease and brain trauma. Increased CD40-CD40 ligation has been implicated in the pathogenesis of several autoimmune inflammatory diseases, such as MS, arthritis and insulin-dependant diabetes.

  • For Western Blot

Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig, rabbit and sheep CD40. Detects a band of ~45kDa by Western blot.

Type: Primary
Antigen: CD40
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty