Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-PK004-D KAP-PK004-F
89141-918EA 499 USD InStock
89141-918 89141-916
Anti-AKT Rabbit Polyclonal Antibody
Antibodies
The Akt (PKB) family of protein kinases are serine/threonine kinases, with three mammalian family members identified (Akt1, Akt2, Akt3). Akt is a well-characterized member of PI3 kinase-mediated signaling pathways, regulating cell growth, apoptosis, glycogen synthesis, and other cellular responses through its phosphorylation of downstream substrates. Akt activation is triggered by binding of phospholipid and phosphorylation at two key residues: Thr308 by PDK1, and Ser473 by PDK2, now identified as mTOR. Deregulation of Akt signaling has been associated with cancer, diabetes, and schizophrenia. Akt1 is the cellular homologue of the murine thymoma retroviral oncogene v-akt, and its role in anti-apoptotic and pro-mitotic pathways have made Akt a molecular target for anti-cancer therapeutic intervention. Akt activation inhibits apoptosis by phosphorylating the Bcl-2 related protein Bad, and increases p53 degradation by phosphorylating mdm2. Mitotic substrates of Akt include GSK-3β, p21CIP1, and p27KIP1, cell cycle inhibitors negatively regulated by Akt phosphorylation. Akt has been shown to mediate angiogenesis through regulation of thrombospondins, which may cooperate with pro-mitotic and anti-apoptotic functions of Akt to promote tumorigenesis.

  • For Immunoprecipitation, Western Blot

Recognizes human, mouse and rat Akt. Detects a band of ~59kDa by Western blot.

Type: Primary
Antigen: AKT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty