Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CNR2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
210-315-R100
89145-472EA 810 USD InStock
89145-472
Anti-CNR2 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human and rat CB2. By Western blot detects bands of ~60kDa from human spinal cord homogenate and ~40kDa from RBL2H3 cells, representing different glycosylated forms of CB2.

Type: Primary
Antigen: CNR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty