Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P2X7 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
215-035-R100
89146-492EA 380 USD InStock
89146-492
Anti-P2X7 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunocytochemistry, Immunoprecipitation

Recognizes mouse purinergic receptor P2X7.

Type: Primary
Antigen: P2X7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty