Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Enhancer for ligands Monoclonal Antibody

Supplier: Enzo Life Sciences
804-034-C050
89153-564EA 399 USD InStock
89153-564
Anti-Enhancer for ligands Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Enhancer for Ligands
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty