Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Apurinic/Apyrimidinic Endonuclease Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
804-213-R100
89153-866EA 842 USD InStock
89153-866
Anti-Apurinic/Apyrimidinic Endonuclease Mouse Monoclonal Antibody
Antibodies
Mammalian apurinic/apyrimidinic (AP) endonuclease (APE1; APEX; HAP1; Ref-1) is a multifunctional, bipartite enzyme that belongs to the class II AP endonucleases and that plays an important role in numerous, cellular functions, like repairing abasic sites (loss of purine or pyrimidine base) in DNA, regulating the redox state of other proteins that play roles in oxidative signalling, transcription factor regulation (Fos, Jun, NF-κ B, Myb, HIF-1α, CREB, Pac), cell cycle control (p53) and apoptosis. APE1 initiates the DNA base excision repair (BER) by cleaving the DNA immediately adjacent to the 5’ of an AP site to produce a hydroxyl group at the 3’ terminus of an unmodified nucleotide upstream of the nick and a 5’-deoxyribose phosphate moiety downstream. The product of APE1 is further processed by DNA polymerase β to release 5’-deoxyribose phosphate with its intrinsic lyase activity. The nicked DNA is then sealed by DNA ligase I or DNA ligaseIII/XRCC1 to complete this repair process. In addition to the AP endonuclease activity, APE1 also possesses a 3’-5’ DNA exonuclease activity, a 3’-phosphodiesterase activity, a 3’-phosphatase activity, and a RNase H activity. It has been shown that APE1 is capable of excision L-configuration deoxyribonucleoside analogs from the 3’ termini of DNA. Human APE1 gene is located on chromosome 14q 11.2-12.

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human APE1. Detects a band of ~37kDa by Western blot.

Type: Primary
Antigen: APURINIC/APYRIMIDINIC ENDONUCLEASE
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty