Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP8 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-447-C100
89154-302EA 406 USD InStock
89154-302
Anti-CASP8 Rat Monoclonal Antibody
Antibodies
Host: Rat, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunocytochemistry, Western Blot

Recognizes the p18 subunit of mouse caspase-8. Detects bands of ~55kDa (full-length caspase-8) and ~18kDa (apoptosis-induced cleavage fragment) by Western blot. Does not cross-react with human caspase-8.

Type: Primary
Antigen: CASP8
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty