Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP1 Rat Monoclonal Antibody

Supplier: Enzo Life Sciences
804-531-C100
89154-482EA 491 USD InStock
89154-482
Anti-CASP1 Rat Monoclonal Antibody
Antibodies
Capases are a highly conserved family of cysteine proteases and are synthesized as inactive zymogens, which are cleaved at Asp-x sites during apoptosis, generating large and small subunits that heterodimerize to form the active enzyme. Caspase-1 is a member of the ICE/CED-3 family of caspases and is detected in the pro-form as a protein of 45 kDa.
Caspase-1 generates the bioactive form of the pro-inflammatory cytokine IL-1β from its biologically inactive precursor. Overexpression of caspase-1 in cells induces apoptosis, which can be inhibited by cytokine response modifier A (CrmA). Caspase-1 mRNA is predominantly expressed in the spleen and lung and to a lesser extent in skeletal muscle, kidney, testis and heart and within these tissues macrophages preferentially express caspase-1. Caspase-1 deficient mice have no gross abnormalities, however, macrophages from caspase-1-deficient mice are defective in IL-β processing and release.

  • For ELISA, Flow Cytometry, Western Blot

Recognizes an epitope within the p20 fragment of mouse caspase-1. Detects a band of ~45kDa by Western blot. Does not cross-react with mouse caspase-11 or -12.

Type: Primary
Antigen: CASP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty