Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LAG3 Mouse Monoclonal Antibody (ATTO 488)

Supplier: Enzo Life Sciences
804-806TD-T100
89155-012EA 729 USD InStock
89155-012
Anti-LAG3 Mouse Monoclonal Antibody (ATTO 488)
Antibodies
The lymphocyte activation gene-3 (LAG-3, CD223), a member of the immunoglobulin superfamily (IgSF) related to CD4, binds to the major histocompatibility complex (MHC) class II molecules but with higher affinity than CD4. Several alternative mRNA splice-variants of human LAG-3 have been described, two of them encoding potential secreted forms: LAG-3V1 (i.e. the D1-D2 domains of the protein, 36 kDa) and LAG-3V3 (D1-D3, 52 kDa). The longer form was detected by ELISA in the serum of healthy individuals as well as of tuberculosis patients with a favorable outcome. LAG-3 expression by T cell clones correlated with IFN-γ production, and hence soluble LAG-3 has been suggested as a serological marker of Th1 responses.

  • For Immunocytochemistry

Recognizes the 30 aa extra-loop of the first N-terminal D1 domain of human LAG-3.

Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone:
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty