Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CEACAM5 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
805-077-C100
89155-366EA 465 USD InStock
89155-366
Anti-CEACAM5 Mouse Monoclonal Antibody
Antibodies
CD66e (CEACAM5; CEA) belongs to the carcinoembryonic antigen (CEA) gene family. It encodes a glycosyl phosphatidyl inositol (GPI)-linked glycoprotein with a Mr of 180'000-200'000 which is most strongly expressed on epithelial cells of the fetal and adult colon and to a minor extent on epithelial cells of the stomach and sweat glands, squamous epithelial cells of the tongue, esophagus and cervix. CD66e is used as a tumor marker for early detection of recurrent disease due to its expression in adenocarcinomas of the colon, lung, breast, stomach and pancreas and in mucinous ovarian carcinomas. Like all members of the CEACAM family, it consists of a single N domain, with structural homology to the immunoglobulin variable domains, followed by six immunoglobulin constant-like A (A1, A2, A3) and B domains (B1, B2, B3).

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes human CD66e. Does not cross-react with human CEACAM1, 3, 4, 6, 7, 8 or PSG.

Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty