Would you like to log in again?
CV-Cathepsin B Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK125-0001
89156-440EA 366 USD InStock
89156-440
CV-Cathepsin B Detection Kit, Enzo Life Sciences
Assays Enzyme Assays
Based on the dye cresyl violet and the cathepsin B ArgArg peptide recognition sequence, this kit allows the detection of cathepsin B activity in living cells by fluorescence microscopy.
Once cells are grown, staining is completed in just a few minutes
Elevated cathepsin enzyme activity often indicates the presence of a pathological condition including cancer, Alzheimer's disease, autoimmune disease, and osteoporosis.
Hoechst 33342 stain and Acridine Orange are also included to allow visualization of nuclei and lysosomes.
Ordering information: Deliver Information: Kit contains CV-(RR)2 reagent, Acridine Orange, Hoechst 33342 stain
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty