Would you like to log in again?
DPPIV Drug Discovery Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK499-0001
89156-500EA 464 USD InStock
89156-500
DPPIV Drug Discovery Kit, Enzo Life Sciences
Assays Enzyme Assays
The DPPIV Drug Discovery Kit is a complete assay system designed to screen DPPIV inhibitors, a potential therapeutic target, providing enough material to perform at least 96 assays.
DPPIV (DPP4, CD26) is a member of the class of proteases known as prolyl peptidases, which cleave proteins after proline residues and is thought to play roles in diabetes, cancer, and autoimmune diseases, making it a target for drug discovery.
The kit contains both a chromogenic substrate (H-Gly-Pro-pNA; Km=114 µM) and a fluorogenic substrate (H-Gly-Pro-AMC; Km=50 µM).
Cleavage of the p-nitroaniline (pNA) from the colorimetric substrate increases absorbance at 405 nm. The fluorimetric assay is based on the cleavage of 7-amino-4-methylcoumarin (AMC) moiety from the C-terminus of the peptide substrate, which increases its fluorescence intensity at 460 nm.
A DPPIV inhibitor, P32/98 (KI=130 nM12), is included for use as a control. Other DPP enzymes are available for specificity profiling.
Ordering information: Contents:
DPPIV enzyme (human, recombinant), pNA substrate, Calibration standard (p-nitroaniline), AMC substrate, Calibration standard (7-Amino-4-methylcoumarin), Inhibitor, Assay Buffer, ½-volume clear microplate
Delivery information: Kit includes DPPIV enzyme (human, recombinant), pNA substrate, Calibration standard (p-nitroaniline), AMC substrate, Calibration standard (7-Amino-4-methylcoumarin), Inhibitor, Assay Buffer, ½-volume clear microplate
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty