Would you like to log in again?
Caspase-3 Cell Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BML-AK703-0001
89156-540EA 579 USD InStock
89156-540
Caspase-3 Cell Assay Kit, Enzo Life Sciences
Assays Cellular Assays
This kit is a complete assay system for measuring caspase-3 and caspase-3-like activity (DEVDase) in cellular extracts.
It is performed in a 96-well format with all reagents for preparing cell extracts, measuring caspase activity and calibrating the assay. The caspase-3, is useful as a positive control, for testing cellular extracts for endogenous inhibitors, and more
Delivery information: Kit includes Caspase-3 enzyme (human, recombinant), pNA Substrate, pNA calibration standard, AMC Substrate, AMC calibration standard (7-amino-4-methylcoumarin), Inhibitor (Ac-DEvD-CHO), Assay buffer, Cell lysis buffer, Half volume microplate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty