Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA266-0020
89162-802EA 204 USD InStock
89162-802
Anti-MAPK Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes human, mouse, rat, dog and carp p38 isoforms p38alpha, p38beta, and p38gamma phosphorylated dually at Thr180 and Tyr182. Does not cross-react with unphosphorylated forms of p38.

Type: Primary
Antigen: MAPK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty