Would you like to log in again?
Patient Control Slide, Azer Scientific
Supplier: Azer Scientific
ESCS-100 ESCS-101
89176-820CS 1242.14 USD InStock
89176-820 89176-822
Patient Control Slide, Azer Scientific
Slides Microscopy Control Slides
The patient control slide is suitable for staining protocols that require a control.
  • Spaces for Both Patient Sample and Control Material
  • Chemical Adhesive for Better Tissue Adhesion
  • Red Line on Reverse Side of Slide for Unrestricted Movement of Reagents
Each slide features space for both patient sample and control material
Slide is positively charged for better tissue adhesions. Available without clipped corners or with clipped corners for use in automated systems.
The slide is positively charged for better tissue adhesions. Available without clipped corners or with clipped corners for use in automated systems.
A red line allows for the two samples to be separated. This red line is printed on reverse side of slide to allow for unrestricted movement of reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty