Would you like to log in again?
Nunc® TufRol Roller Bottles, PS, Thermo Scientific
Supplier: Thermo Scientific
184344 184944 184344 184902 182702 184902 142720 141744 184920 182744 181702 184944 184302 181702 184302 182720 184920 142744 183302 182744 182720 141744 182702 183302 183902 142744 183902 142720
89203-536CS 262.38 USD InStock
89203-536 CA89203-522 89203-534 CA89203-542 CA89203-520 89203-532 CA89203-540 89203-530 CA89218-770 CA89203-518 CA89203-538 89203-528 CA89203-536 89203-526 CA89203-534 89218-770 89203-524 CA89203-532 89203-522 CA89203-530 89203-542 89203-520 89203-540 CA89203-528 89203-518 CA89203-526 89203-538 CA89203-524
Nunc® TufRol Roller Bottles, PS, Thermo Scientific
Bottles Roller Bottles
Nunc® Tufrol PS roller bottles have been developed for applications such as industrial scale production of vaccines, monoclonal antibodies, and biotherapeutics.
Bottles provide the reliability and reproducibility necessary for cell growth needs. The one-piece seamless design reduces the possibility of leakage through seams.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty